Learn More
Medchemexpress LLC Alkali lignin | 8068-05-1 | MFCD09039274 | >98.0% | No single chemical formula - complex, polydisperse polymer | 25 G
SDP
Catalog No. 5000281296
Encompass_Preferred
Lignin alkali is an alkali-extracted lignin supplied as a research-grade biochemical reagent in powder form. It serves as an organic material or reagent for life-science and materials research, including biochemical assays, polymer studies, and material formulation investigations.
  • High purity: ≥98.0%.
  • Solid powder, brown to black in color.
  • Suitable as a biochemical reagent or organic material for research applications.
  • Available in multiple pack sizes, including 25 G, for lab-scale studies.
  • CAS number: 8068-05-1.
  • Store sealed, away from moisture at 4°C; in solvent store at -80°C (long term) or -20°C (short term).
Catalog No. 50-002-81296 Supplier Medchemexpress LLC Supplier No. HYW25011225G
Lignin alkali is an alkali-extracted lignin supplied as a research-grade biochemical reagent in powder form. It serves as an organic material or reagent for life-science and materials research, including biochemical assays, polymer studies, and material formulation investigations.
  • High purity: ≥98.0%.
  • Solid powder, brown to black in color.
  • Suitable as a biochemical reagent or organic material for research applications.
  • Available in multiple pack sizes, including 25 G, for lab-scale studies.
  • CAS number: 8068-05-1.
  • Store sealed, away from moisture at 4°C; in solvent store at -80°C (long term) or -20°C (short term).