Learn More
Alkali Scientific Alkali Scientific™ Analytical Balance with Quick-Cal™ internal calibration, 120 grams
SDP
Catalog No. 502335959
Encompass_Preferred
• Readablity to 0.0001g (0.1 mg)• Available with Quick-Cal™ internal calibration feature• RS232 port for data output to a computer or printer• Multiple mode options (standard weighing, count mode, & percent)Accuris Analytical balances incorporate electromagnetic sensor technology and precision mechanical components to offer superior weight determinations to a fraction of a milligram. Two capacity ranges are available in the Analytical Balance line to weigh samples up to 120g or 210g. Readability for both ranges is 0.0001g (0.1mg), with accuracy and linearity specifications that match up with the top brands in the industry. The W3100 balances tare and stabilize quickly to provide immediate results.Both ranges offer an optional “Quick-CalTM” internal calibration feature that allows quick and convenient calibration without the need for a calibration standard weight. The glass draft shield enclosing the weighing chamber has sliding doors on the sides and top for easy acces
Catalog No. 50-233-5959 Supplier Alkali Scientific Supplier No. W3100A120
• Readablity to 0.0001g (0.1 mg)• Available with Quick-Cal™ internal calibration feature• RS232 port for data output to a computer or printer• Multiple mode options (standard weighing, count mode, & percent)Accuris Analytical balances incorporate electromagnetic sensor technology and precision mechanical components to offer superior weight determinations to a fraction of a milligram. Two capacity ranges are available in the Analytical Balance line to weigh samples up to 120g or 210g. Readability for both ranges is 0.0001g (0.1mg), with accuracy and linearity specifications that match up with the top brands in the industry. The W3100 balances tare and stabilize quickly to provide immediate results.Both ranges offer an optional “Quick-CalTM” internal calibration feature that allows quick and convenient calibration without the need for a calibration standard weight. The glass draft shield enclosing the weighing chamber has sliding doors on the sides and top for easy acces