Learn More
Alkali Scientific Alkali Scientific™ Compact Balance, 5000 grams
SDP
Supplier:  Alkali Scientific W33005000
Encompass_Preferred
Compact housing: 7.5 x 11 inch footprint • Readablity 0.1 to 0.01g • Easy in-lab calibration feature • RS232 port for data output to a computer or printer • Multiple mode options (standard weighing, count mode, & percent) • Simple, reliable performanceAccuris Compact balances have been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0.01g to 0.1 g. Their reliable and precise weighing technology utilizes gold-plated, ceramic capacitors to determine sample mass. The Compact line includes 6 models covering a wide range of maximum weight capacities. The lower capacity units (120g, 300g, and 500g) are readable to 0.01 g, and come with a draft shield ring to prevent air currents from affecting measurements. These models feature a 4.5 inch diameter, stainless steel weigh pan. The larger capacity models (1000g, 5000g, and 10,000g) do not include the draft shield ring, come with larger weighing pans of 6.3 to 7 inch dia
Catalog No. 50-233-6083