Learn More
Alkali Scientific CellPro™ 1.6 ml 96-Well Deep Well Plate, U-Bottom, Square Well, 5/pk, 50/cs
SDP
Catalog No. 502388966
Encompass_Preferred
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 50-238-8966 Supplier Alkali Scientific Supplier No. MPD24
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).