Learn More
Alkali Scientific CellPro™ 2.2 ml 96-Well Deep Well Plate, U-Bottom, Square Well, 5/pk, 50/cs
SDP
Catalog No. 502388819
Encompass_Preferred
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 50-238-8819 Supplier Alkali Scientific Supplier No. MPD44
Add to Cart
Add to Cart
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).