Learn More
Alkali Scientific Alkali Scientific™ CO2 Incubator,180 Liter, 115V with two shelves. Includes: High heat decontamination, humidity control and split inner door with 6 compartments
SDP
Catalog No. 502388911
Encompass_Preferred
Alkali Scientific™ CO2 Incubator,180 Liter, 115V with two shelves. Includes: High heat decontamination, humidity control and split inner door with 6 compartments
Catalog No. 50-238-8911 Supplier Alkali Scientific Supplier No. RS8093
Add to Cart
Add to Cart
Request a Quote
Alkali Scientific™ CO2 Incubator,180 Liter, 115V with two shelves. Includes: High heat decontamination, humidity control and split inner door with 6 compartments