Learn More
Hach Company Alkali Solution for Calcium and Magnesium, 100 mL MDB
Supplier:  Hach Company 2241732
Encompass
For determination of calcium and magnesium hardness in water by the Calmagite Colorimetric method. Hach Method 8030. 100 mL Marked Dropping Bottle.
Catalog No. 50-199-5602