Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coast Alkaline Battery 9 V Industrial Performance

Catalog No. 17018113
Change view
Click to view available options
Quantity:
12/Pk.
4/Pk.
48/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-018-113 4/Pk.
17-018-044 12/Pk.
17-004-841 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-018-113 Supplier Coast Supplier No. 30602
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to maximize the performance of all types of handheld/headlamp devices, COAST Industrial Performance (IP) 9V batteries deliver top-rated power7mdash; whether you’re on the worksite, at home, or on the go

  • Remove the stress from critical situations and simplify routine, practical situations by choosing batteries that take performance to the next level
  • When you choose COAST Industrial Performance batteries, you get the quality guarantee of a company that’s been at it for 100 years—with products that make people’s work safer and lives easier

Specifications

Product Type Alkaline Battery
Battery Type 9 V
Material Alkaline Manganese
Cell Type Button
Quantity 4/Pk.
Voltage 9 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.