Learn More
Alkaline Blue, 0.04% in Methanol, Reagents
Specifications
Specifications
| Chemical Name or Material | Alkaline Blue, 0.04% in Methanol |
| CAS | 67-56-1, 1324-76-1 |
| Boiling Point | 65°C |
| Packaging | Glass Bottle |
| Specific Gravity | 0.7932 |
| Color | Blue |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Percent Purity | 0.04% |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.