Learn More
Bioworld Alkaline Buffer, 1.5M, pH 10.3, 100 mL
Catalog No. 50255343
Encompass_Preferred
Product Description Alkaline buffer has a 1.5M concentration and a pH level of 10.3. It contains 2-amino-2-methylpropanol. This buffer is used in alkaline phosphatase reactions in conjunction with p-nitrophenyl phosphate (pNPP) enzyme substrate.
Catalog No. 50-255-343 Supplier Bioworld Supplier No. 401200611
Product Description Alkaline buffer has a 1.5M concentration and a pH level of 10.3. It contains 2-amino-2-methylpropanol. This buffer is used in alkaline phosphatase reactions in conjunction with p-nitrophenyl phosphate (pNPP) enzyme substrate.