Learn More
Hach Company Alkaline Cyanide Reagent, 100 mL MDB
Supplier:  Hach Company 2122332
Encompass
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn. 100 mL Marked Dropping Bottle.
Catalog No. 50-199-5450