Learn More
MSC ZEP FS Process Cleaner, Liquid, Alkaline, 1 gal
Catalog No. 502818491
Encompass
ZEP FS Process Cleaner is an alkaline all-purpose cleaner and degreaser designed for use in food and beverage environments. It is a clear, green liquid with a mild odor and a high pH for effective cleaning.
  • Effectively cleans and degreases surfaces
  • Alkaline formula with a pH range of 13.0–13.5
  • Soluble in both cold and hot water
  • Recommended for food and beverage applications
Catalog No. 50-281-8491 Supplier MSC Supplier No. 17641093
ZEP FS Process Cleaner is an alkaline all-purpose cleaner and degreaser designed for use in food and beverage environments. It is a clear, green liquid with a mild odor and a high pH for effective cleaning.
  • Effectively cleans and degreases surfaces
  • Alkaline formula with a pH range of 13.0–13.5
  • Soluble in both cold and hot water
  • Recommended for food and beverage applications