Learn More
Bioworld Alkaline DNA Gel Running Buffer, 250 mL
Catalog No. 50196650
Encompass_Preferred
Product DescriptionAlkaline DNA gel running buffer contains 0.5M NaOH and 50 mM EDTA. EDTA is to chelate divalent metal cations such as Mg2+ and Ca2+, which are required for DNA function of degrading enzymes (DNAses).
Catalog No. 50-196-650 Supplier Bioworld Supplier No. 105300001
Product DescriptionAlkaline DNA gel running buffer contains 0.5M NaOH and 50 mM EDTA. EDTA is to chelate divalent metal cations such as Mg2+ and Ca2+, which are required for DNA function of degrading enzymes (DNAses).