Learn More
Hach Company Alkaline Iodide-Azide Reagent, 500 mL
Supplier:  Hach Company 27749
Encompass
For determination of Dissolved Oxygen by the Winkler Method. APHA formulation
Catalog No. 50-199-4230