Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkaline Phosphatase
Catalog No. 502900596
Encompass
Alkaline phosphatase catalyzes the removal of phosphate group from various compounds that are phosphorylated. Alkaline phosphatase from calf intestine hydrolyzes 5AE-monophosphate groups from both DNA and RNA. It can also hydrolyze 5AE-diphosphate and 5AE-triphosphate groups from RNA.
Catalog No. 50-290-0596 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10713023001
Alkaline phosphatase catalyzes the removal of phosphate group from various compounds that are phosphorylated. Alkaline phosphatase from calf intestine hydrolyzes 5AE-monophosphate groups from both DNA and RNA. It can also hydrolyze 5AE-diphosphate and 5AE-triphosphate groups from RNA.