Learn More
Bioworld Alkaline Phosphatase, 2500 Unit(s)
Catalog No. 50198957
Encompass_Preferred
Alkaline Phosphatase1,000 UConjugation GradeAmmonium Sulfate SuspensionSource: Calf Intestine Adenosine Deaminase: <=0.001%Concentration (37 C): >7500U/mlPhosphodiesterase: <=0.001%DNase, RNase, and Protease: NoneSpecific activity (37 C, pH 10.5, DEA): >1500U/mg
Catalog No. 50-198-957 Supplier Bioworld Supplier No. 215600002
Alkaline Phosphatase1,000 UConjugation GradeAmmonium Sulfate SuspensionSource: Calf Intestine Adenosine Deaminase: <=0.001%Concentration (37 C): >7500U/mlPhosphodiesterase: <=0.001%DNase, RNase, and Protease: NoneSpecific activity (37 C, pH 10.5, DEA): >1500U/mg