Learn More
Bioworld Alkaline Phosphatase in 50% Glycerol, 1000 Unit(s)
Catalog No. 50198958
Encompass_Preferred
Alkaline Phosphatase in 50% GlycerolSpecifications:Source: Calf Intestine.Supplied in 50% GlycerolActivity: 3000 - 4000 U/mg proteinApplication: as an indicator enzyme when conjugted to proteins such as antibodies, avidin etc. Suitable also for western blot analysis.Unit definition: the amount of enzyme catalyzing the liberation of 1 μmole of p-nitrophenyl phosphate per minute at 37° C, pH 9.8 in diethanolamine buffer, 405nm.If assayed at 25°C in glycine buffer, pH 9.6, then, 1 glycine unit= 3DEA units
Catalog No. 50-198-958 Supplier Bioworld Supplier No. 215600011
Alkaline Phosphatase in 50% GlycerolSpecifications:Source: Calf Intestine.Supplied in 50% GlycerolActivity: 3000 - 4000 U/mg proteinApplication: as an indicator enzyme when conjugted to proteins such as antibodies, avidin etc. Suitable also for western blot analysis.Unit definition: the amount of enzyme catalyzing the liberation of 1 μmole of p-nitrophenyl phosphate per minute at 37° C, pH 9.8 in diethanolamine buffer, 405nm.If assayed at 25°C in glycine buffer, pH 9.6, then, 1 glycine unit= 3DEA units