Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphatase, Alkaline from bovine intestinal mucosa, 9001-78-9, MFCD00131849, 500 mg
Catalog No. NC1100499
Encompass
Lyophilized powder, ≥10 DEA units/mg solid, storage temp: −20°C, Synonym: Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum).
Catalog No. NC1100499 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P7640500MG
Lyophilized powder, ≥10 DEA units/mg solid, storage temp: −20°C, Synonym: Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum).