Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkaline Phosphatase AP gr
Catalog No. 502900512
Encompass
Orthophosphoric-monoester phosphohydrolase (alkaline optimum)Alkaline phosphatase catalyzes the removal of phosphate group from various compounds that are phosphorylated. It hydrolyzes 5AE-monophosphate groups from both DNA and RNA. It can also hydrolyze 5AE-diphosphate and 5AE-triphosphate groups from RNA.
Catalog No. 50-290-0512 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10108138001
Orthophosphoric-monoester phosphohydrolase (alkaline optimum)Alkaline phosphatase catalyzes the removal of phosphate group from various compounds that are phosphorylated. It hydrolyzes 5AE-monophosphate groups from both DNA and RNA. It can also hydrolyze 5AE-diphosphate and 5AE-triphosphate groups from RNA.