Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alkaline phosphatase (Calf), EIA & Molecular Biology Grade

Catalog No. 89933182 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-182 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-182 Supplier Abnova™ Supplier No. P5282
Only null left
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Concentration ∼ 20 mg/mL
For Use With (Application) Enzyme Activity
Formulation In 50% glycerol containing 5mM MgCl
Molecular Weight (g/mol) 140kDa
Name Alkaline phosphatase (Calf), EIA & Molecular Biology Grade
Preparation Method Native protein chromatographically purified from Calf Intestine
Quantity 1 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name ALKALINE
Assays Alkaline Phosphatase Assay
Biological Activity ≥3,000 units per mg protein (37°C, pH9.8, DEA). One Unit hydrolyzes 1 umole of p-nitrophenol phosphate per minute at 37°C, pH 9.8.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Calf Intestine
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.