Learn More
Abcam Alkaline phosphatase Conjugation Kit - Lightning-Link, 100UG
Catalog No. 502585623
Encompass_Preferred
Abcam's Alkaline Phosphatase Conjugation Kit (ab102850) offers : Rapid Conjugation: Achieve alkaline phosphatase labeling in under 4 hours with just 30 seconds of hands-on time. High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5623 Supplier Abcam Supplier No. AB102850100UG
Abcam's Alkaline Phosphatase Conjugation Kit (ab102850) offers : Rapid Conjugation: Achieve alkaline phosphatase labeling in under 4 hours with just 30 seconds of hands-on time. High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement