Learn More
Abcam Alkaline Phosphatase Conjugation Kit - Lightning-Link, 3X100 μg, for antibody and protein labeling, 30 seconds hands-on time, 100% antibody recovery
Catalog No. NC3306257
Encompass_Preferred
The Abcam Alkaline Phosphatase Conjugation Kit - Lightning-Link offers a rapid and efficient method for covalent labeling of various biomolecules containing primary amines. This easy-to-use kit requires only 30 seconds of hands-on time and produces conjugates ready for use in less than 4 hours without the need for purification, ensuring 100% antibody recovery. Designed specifically for applications in immunology, it is suitable for Western blotting, ELISA, and immunohistochemistry. Its recombinant alkaline phosphatase enzyme ensures consistent performance across batches.
Catalog No. NC3306257 Supplier Abcam Supplier No. AB1028503X100UG
The Abcam Alkaline Phosphatase Conjugation Kit - Lightning-Link offers a rapid and efficient method for covalent labeling of various biomolecules containing primary amines. This easy-to-use kit requires only 30 seconds of hands-on time and produces conjugates ready for use in less than 4 hours without the need for purification, ensuring 100% antibody recovery. Designed specifically for applications in immunology, it is suitable for Western blotting, ELISA, and immunohistochemistry. Its recombinant alkaline phosphatase enzyme ensures consistent performance across batches.