Learn More
Abcam Alkaline phosphatase Conjugation Kit - Lightning-Link, 3 x 10UG
Catalog No. 502585621
Encompass_Preferred
Abcam's Alkaline Phosphatase Conjugation Kit (ab102850) offers : Rapid Conjugation: Achieve alkaline phosphatase labeling in under 4 hours with just 30 seconds of hands-on time. High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5621 Supplier Abcam Supplier No. AB1028503X10UG
Abcam's Alkaline Phosphatase Conjugation Kit (ab102850) offers : Rapid Conjugation: Achieve alkaline phosphatase labeling in under 4 hours with just 30 seconds of hands-on time. High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement