Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkaline Phosphatase Dieth
Catalog No. 502905126
Encompass
The Alkaline Phosphatase Diethanolamine Detection Kit provides ready-to-use reagents for detecting the presence of alkaline phosphatase activity. This simple assay to detect alkaline phosphatase activity uses p-Nitrophenyl Phosphate (pNPP) as the substrate.
Catalog No. 50-290-5126 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. AP01001KT
The Alkaline Phosphatase Diethanolamine Detection Kit provides ready-to-use reagents for detecting the presence of alkaline phosphatase activity. This simple assay to detect alkaline phosphatase activity uses p-Nitrophenyl Phosphate (pNPP) as the substrate.