Learn More
Crescent Chemical Co Inc ALKALINE PHOSPHATASE FROM CALF
SDP
Catalog No. 50248013
Encompass_Preferred
Alkaline Phosphatase from calf intestine ca. 3000 U/mg protein solution Especially suitable for the preparation of EIA-conjugates. Further dialysis is unnecessary (1). In 50 % glycerol, containing 5 mM Tris, 5 mM MgCl2, 0.1 mM ZnCl2, pH ca. 7.0. Size - 5MG Storage Conditions - +2 °C TO +8 °C Ca
Catalog No. 50-248-013 Supplier Crescent Chemical Co Inc Supplier No. SE32471.02
Alkaline Phosphatase from calf intestine ca. 3000 U/mg protein solution Especially suitable for the preparation of EIA-conjugates. Further dialysis is unnecessary (1). In 50 % glycerol, containing 5 mM Tris, 5 mM MgCl2, 0.1 mM ZnCl2, pH ca. 7.0. Size - 5MG Storage Conditions - +2 °C TO +8 °C Ca