Learn More
Crescent Chemical Co Inc ALKALINE PHOSPHATASE FROM CALF
Supplier: Crescent Chemical Co Inc SE32471.01
Alkaline Phosphatase from calf intestine ca. 3000 U/mg protein solution Especially suitable for the preparation of EIA-conjugates. Further dialysis is unnecessary (1). In 50 % glycerol, containing 5 mM Tris, 5 mM MgCl2, 0.1 mM ZnCl2, pH ca. 7.0. Size - 1MG Storage Conditions - +2 °C TO +8 °C Ca