Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase, MP Biomedicals™

Catalog No. ICN15934705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15934705 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15934705 Supplier MP Biomedicals, Inc Supplier No. 0215934705
Add to Cart
Add to Cart

Specifications

CAS 9001-78-9
Content And Storage Store at 0°C.
Format Solid
Molecular Weight (g/mol) 496.569
Product Type Alkaline Phosphatase
Quantity 5mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.