Learn More
Genscript Corporation Alkaline Phosphatase Placenta
Catalog No. 5000059256
Encompass
Alkaline phosphatase can be considered our favorite enzyme for reasons apparent to those who diagnose and treat metabolic bone diseases or who study skeletal biology Few might know however that alkaline phosphatase likely represents the most frequently assayed enzyme in all of medicine Elevated activity in the circulation is universally recognized as a marker for skeletal or hepatobiliary disease
Catalog No. 50-000-59256 Supplier Genscript Corporation Supplier No. Z04357500
Alkaline phosphatase can be considered our favorite enzyme for reasons apparent to those who diagnose and treat metabolic bone diseases or who study skeletal biology Few might know however that alkaline phosphatase likely represents the most frequently assayed enzyme in all of medicine Elevated activity in the circulation is universally recognized as a marker for skeletal or hepatobiliary disease