Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Alkaline Phosphatase Reagents

Catalog No. 01801136 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
4 x 180 mL Reagent 1, 4 x 180 mL Reagent 2
4 x 30 mL Reagent 1, 4 x 30 mL Reagent 2
4 x 53 mL Reagent 1, 4 x 53 mL Reagent 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-801-136 4 x 30 mL Reagent 1, 4 x 30 mL Reagent 2
01-801-048 4 x 53 mL Reagent 1, 4 x 53 mL Reagent 2
01-801-082 4 x 180 mL Reagent 1, 4 x 180 mL Reagent 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-801-136 Supplier Beckman Coulter™ Supplier No. OSR6104
Add to Cart
Add to Cart
This item is not returnable. View return policy

Alkaline Phosphatase Reagents 1 and 2 for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Includes 4 x 30 mL Reagent 1, 4 x 30 mL Reagent 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.