Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkaline Phosphatase Yello
Catalog No. 502900006
Encompass
The product is ready to use buffered alkaline phosphatase substrate consist of p-nitrophenylphosphate (pNPP). This is the advanced substrate system contains insoluble stabilizer that enhances the stability and reactivity as well as reduces the base-line absorption. It produces a soluble end product hence not recommended for blotting or histochemistry applications.
Catalog No. 50-290-0006 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P7998100ML
The product is ready to use buffered alkaline phosphatase substrate consist of p-nitrophenylphosphate (pNPP). This is the advanced substrate system contains insoluble stabilizer that enhances the stability and reactivity as well as reduces the base-line absorption. It produces a soluble end product hence not recommended for blotting or histochemistry applications.