Learn More
Vector Laboratories VECTASTAIN ABC-AP Staining Kit (Alkaline Phosphatase, Standard)
SDP
Catalog No. NC9175589
Encompass
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories to provide reliable results. All of the VECTASTAIN kits are based on the versatile avidin-biotin complex (ABC) interaction, and different kit variations can be used to accommodate a wide array of primary antibody and tissue species. The avidin-biotin complex, once formed, is stable and can be used for several days after preparation. VECTASTAIN ABC-AP Standard Kit is an option for customers who want to use their own biotinylated secondary antibody. This kit uses alkaline phosphatase (AP) as a detection enzyme. The VECTASTAIN ABC-AP Standard Kit may be used to detect any biotinylated target, not just secondary antibodies. Includes: 3 ml normal horse serum, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated AP). This kit does not include a species-specific secondary antibody. Stains 1000-2000 sections.
Catalog No. NC9175589 Supplier Vector Laboratories Supplier No. AK5000
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories to provide reliable results. All of the VECTASTAIN kits are based on the versatile avidin-biotin complex (ABC) interaction, and different kit variations can be used to accommodate a wide array of primary antibody and tissue species. The avidin-biotin complex, once formed, is stable and can be used for several days after preparation. VECTASTAIN ABC-AP Standard Kit is an option for customers who want to use their own biotinylated secondary antibody. This kit uses alkaline phosphatase (AP) as a detection enzyme. The VECTASTAIN ABC-AP Standard Kit may be used to detect any biotinylated target, not just secondary antibodies. Includes: 3 ml normal horse serum, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated AP). This kit does not include a species-specific secondary antibody. Stains 1000-2000 sections.