Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Alkaline Potassium Iodide-Azide Reagent, 30ml
SDPSCI_ED

Catalog No. S72327 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72327 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72327 Supplier LaMotte™ Supplier No. 7166G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement reagent for LaMotte dissolved oxygen modified Winkler titration test.

  • 30ml bottle
  • Replacement reagent for 5860-01 Dissolved Oxygen Test Kit
  • Additional reagents required to perform the complete test

Specifications

For Use With (Application) 5860-01 Dissolved Oxygen Test Kit
Quantity 30 mL
Range N/A
Type Replacement test kit reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.