Learn More
Ethos Biosciences Sodium Hydroxide 1.0N, 1gal, Ethos
Catalog No. 5000574328
Encompass_Preferred
Sodium Hydroxide 1N is a standardized, ready-to-use alkaline solution formulated for accurate and reliable laboratory applications. Ideal for acid-base titrations, quantitative analysis, pH adjustment, and reagent preparation, this solution is precisely prepared to deliver consistent normality and dependable performance. Manufactured under controlled conditions to support quality and reproducibility, Sodium Hydroxide 1N helps minimize dilution errors and reduce preparation time. It is suitable for use in clinical, research, industrial, and educational laboratory settings where consistent analytical results are essential.
Catalog No. 50-005-74328 Supplier Ethos Biosciences Supplier No. 3261G
Sodium Hydroxide 1N is a standardized, ready-to-use alkaline solution formulated for accurate and reliable laboratory applications. Ideal for acid-base titrations, quantitative analysis, pH adjustment, and reagent preparation, this solution is precisely prepared to deliver consistent normality and dependable performance. Manufactured under controlled conditions to support quality and reproducibility, Sodium Hydroxide 1N helps minimize dilution errors and reduce preparation time. It is suitable for use in clinical, research, industrial, and educational laboratory settings where consistent analytical results are essential.