Learn More
Ethos Biosciences Potassium Hydroxide 20%, 1gal, Ethos
Catalog No. 5000574146
Encompass_Preferred
Potassium Hydroxide 20% is an alkaline solution suitable for microbiology, pathology, cytology, and general laboratory applications. Commonly used for KOH preparations, it is ideal for clearing skin, hair, and nail specimens to aid in the microscopic detection of fungal elements. The clear, colorless solution provides consistent performance for reliable diagnostic and analytical results. It is also appropriate for tissue digestion, pH adjustment, cleaning, and general chemical processing.
Catalog No. 50-005-74146 Supplier Ethos Biosciences Supplier No. 3395G
Potassium Hydroxide 20% is an alkaline solution suitable for microbiology, pathology, cytology, and general laboratory applications. Commonly used for KOH preparations, it is ideal for clearing skin, hair, and nail specimens to aid in the microscopic detection of fungal elements. The clear, colorless solution provides consistent performance for reliable diagnostic and analytical results. It is also appropriate for tissue digestion, pH adjustment, cleaning, and general chemical processing.