Learn More
Ethos Biosciences Potassium Hydroxide 20%, 2oz, Ethos
Catalog No. 5000574343
Encompass_Preferred
Potassium Hydroxide 20% is an alkaline solution suitable for microbiology, pathology, cytology, and general laboratory applications. Commonly used for KOH preparations, it is ideal for clearing skin, hair, and nail specimens to aid in the microscopic detection of fungal elements. The clear, colorless solution provides consistent performance for reliable diagnostic and analytical results. It is also appropriate for tissue digestion, pH adjustment, cleaning, and general chemical processing.
Catalog No. 50-005-74343 Supplier Ethos Biosciences Supplier No. 3395D02
Potassium Hydroxide 20% is an alkaline solution suitable for microbiology, pathology, cytology, and general laboratory applications. Commonly used for KOH preparations, it is ideal for clearing skin, hair, and nail specimens to aid in the microscopic detection of fungal elements. The clear, colorless solution provides consistent performance for reliable diagnostic and analytical results. It is also appropriate for tissue digestion, pH adjustment, cleaning, and general chemical processing.