Description
- Ideal for mobile treatment engineers
- Compact and portable design
- Easy to follow instructions including pictograms
- Complete with all necessary equipment
Specifications
Specifications
| pH Range | 0 to 14 |
| Volume (Metric) | 40 mL |
| No. of Tests | 100 Tests |
| Range | 50 to 2400 mg/L CaCO3 |
| Type | Alkalinity Drop Test |