Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkane Standard Mixture, For Performance Tests of GC-systems, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111019526 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9526 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9526 Supplier MilliporeSigma Supelco Supplier No. 682812MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10 - C40 (all even), 50 mg/L Each

TRUSTED_SUSTAINABILITY

Specifications

Description Contains all even n-alkanes from C10 to C40, 50 mg/L in n-heptane each
Density 0.684 g/mL (at 20°C)
Synonym Determination of hydrocarbons (even-numbered) according to ISO 16703 and EN 14039
Quantity 2 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.