Learn More
Alkane standard Solution C21-C40, Analytical Standard, MilliporeSigma™ Supelco™
Catalog No. 111016489 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6489 5 mL
11-101-6490 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6489 Supplier MilliporeSigma Supelco Supplier No. 040715ML
Only null left
Add to Cart
Add to Cart
Specifications
Description Contains C21, C22, C23, C24, C25, C26, C27, C28, C29, C30, C31, C32, C33, C34, C35, C36, C37, C38, C39, C40, 40 mg/L each, in toluene
Quantity 5 mL
Grade Analytical Standard