Learn More
Alkane standard Solution C8-C20, Analytical Standard, Contains C8-C20, ∼40 mg/L each, in Hexane, MilliporeSigma™ Supelco™
Catalog No. 111020014 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0014 1 mL
11-101-6508 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0014 Supplier MilliporeSigma Supelco Supplier No. 040701ML
Specifications
Description Contains C8, C9, C10, C11, C12, C13, C14, C15, C16, C17, C18, C19, C20; ˜40 mg/L each in hexane
Quantity 1 mL
Grade Analytical Standard