Learn More
ABclonal Technology ALKBH1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A140791000UL
Encompass_Preferred
This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine.
Catalog No. 50-001-06489