Learn More
ABclonal Technology ALKBH1 Rabbit pAb
SDP
Catalog No. 5000106800
Encompass_Preferred
This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine.
Catalog No. 50-001-06800 Supplier ABclonal Technology Supplier No. A14079500UL
This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine.