Learn More
Aviva Systems Biology Corp ALKBH8 Antibody - N-terminal region (ARP41106_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41106_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALKBH8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALKBH8. Alternate names: ABH8, FLJ38204, MGC10235ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-57725