Learn More
Cayman Chemical ALKCD246CTerm RabbIt Mon 100uL
SDP
Catalog No. 502948791
Encompass_Preferred
A catechol estrogen and minor metabolite of estrogen production is increased in microsomes prepared from human mammary adenocarcinoma and fibroadenoma microsomes compared with microsomes prepared from non-cancerous tissues induces renal tumor formation in male hamsters at 25 mg/animal
Catalog No. 50-294-8791 Supplier Cayman Chemical Supplier No. 32287100 UL
A catechol estrogen and minor metabolite of estrogen production is increased in microsomes prepared from human mammary adenocarcinoma and fibroadenoma microsomes compared with microsomes prepared from non-cancerous tissues induces renal tumor formation in male hamsters at 25 mg/animal