Learn More
MSC Detco Cleaner and Degreaser, Liquid, Butyl-Based, Non-Acid, 5 gal
Catalog No. 502816281
Encompass
This Detco cleaner and degreaser is a liquid, butyl-based concentrate designed for various washable surfaces. It is non-acidic and biodegradable, offering effective cleaning in a dilutable formula.
  • Liquid form
  • Butyl-based formula
  • No scent
  • Container size: 5 gal drum
  • Dilutable concentrate solution
  • Non-acidic
  • Biodegradable
  • VOC content 52 g/L
Catalog No. 50-281-6281 Supplier MSC Supplier No. 33701434
This Detco cleaner and degreaser is a liquid, butyl-based concentrate designed for various washable surfaces. It is non-acidic and biodegradable, offering effective cleaning in a dilutable formula.
  • Liquid form
  • Butyl-based formula
  • No scent
  • Container size: 5 gal drum
  • Dilutable concentrate solution
  • Non-acidic
  • Biodegradable
  • VOC content 52 g/L