Learn More
MSC Detco Cleaner and Degreaser, Liquid, Butyl-Based, Unscented, 330 gal
Catalog No. 502819779
Encompass
This Detco Cleaner & Degreaser is a liquid, butyl-based formula, supplied in a 330-gallon tote. It is designed for effective cleaning on washable surfaces and is biodegradable, offering an unscented solution.
  • Liquid, butyl-based formula
  • Supplied in a 330 gal tote container
  • Unscented and biodegradable
  • Dilutable concentrate for various cleaning tasks
  • Non-acidic with a VOC content of 52 g/L (5%)
  • Suitable for use on washable surfaces
Catalog No. 50-281-9779 Supplier MSC Supplier No. 33701475
This Detco Cleaner & Degreaser is a liquid, butyl-based formula, supplied in a 330-gallon tote. It is designed for effective cleaning on washable surfaces and is biodegradable, offering an unscented solution.
  • Liquid, butyl-based formula
  • Supplied in a 330 gal tote container
  • Unscented and biodegradable
  • Dilutable concentrate for various cleaning tasks
  • Non-acidic with a VOC content of 52 g/L (5%)
  • Suitable for use on washable surfaces