Learn More
MSC Detco Cleaner and Degreaser, Liquid, Butyl-Based, Unscented, 55 gal
Catalog No. 502866578
Encompass
This biodegradable cleaner and degreaser is supplied as a liquid in a 55 gal drum. Its butyl-based and non-acidic formula is designed for washable surfaces, offering a dilutable concentrate with no scent.
  • Liquid form for versatile application
  • Biodegradable for environmental consideration
  • Butyl-based formula for effective cleaning
  • Unscented for sensitive environments
  • Non-acidic formulation
  • Dilutable concentrate for customizable strength
  • Suitable for washable surfaces
  • Container type: Drum
  • Container size: 55 gal
  • VOC Content: 52 g/L (5%)
Catalog No. 50-286-6578 Supplier MSC Supplier No. 33701467
This biodegradable cleaner and degreaser is supplied as a liquid in a 55 gal drum. Its butyl-based and non-acidic formula is designed for washable surfaces, offering a dilutable concentrate with no scent.
  • Liquid form for versatile application
  • Biodegradable for environmental consideration
  • Butyl-based formula for effective cleaning
  • Unscented for sensitive environments
  • Non-acidic formulation
  • Dilutable concentrate for customizable strength
  • Suitable for washable surfaces
  • Container type: Drum
  • Container size: 55 gal
  • VOC Content: 52 g/L (5%)