Learn More
Sigma Aldrich Alkyl Naphthalene Sulfonate Sodium Salt
Catalog No. 916994500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
500 g
Catalog No. Quantity
91-699-4500G 500 g
1 options
Catalog No. 91-699-4500G Supplier Sigma Aldrich Supplier No. 916994500G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
This is an ionic surfactant mixture. It provides excellent wetting, dispersing, hydrotroping and medium to low foaming. It offers acid and base stability, hard-water tolerance and high temperature stability.
Specifications
Quantity 500 g
Synonym Alkylated naphthalene sulfonic acid sodium salt; Sodium alkylnapthalene sulfonate