Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkylphenol Internal Standard Mix 7-Solution for DIN EN ISO 18857-2 in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111016611 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6611 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6611 Supplier MilliporeSigma Supelco Supplier No. 3362710ML
Add to Cart
Add to Cart

Certan Vial

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1
Flash Point -17°C - Closed Cup
Type Analytical Standard
Quantity 10 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.