Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkyltrimethylammonium bro500G
Catalog No. 502904498
Encompass
Alkyltrimethylammonium bromide is a surfactant and can be used to study the forces and stability of foam films produced from soluble cationic surfactants.
Catalog No. 50-290-4498 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M7635500G
Alkyltrimethylammonium bromide is a surfactant and can be used to study the forces and stability of foam films produced from soluble cationic surfactants.