Learn More
Medchemexpress LLC Alkyne-βAG-TOCA | >98.99% | 1100.22 | 5 MG
SDP
Catalog No. 5000424987
Encompass_Preferred
Alkyne-βAG-TOCA is an octreotide derivative that targets somatostatin receptor type 2 (SST2). This compound is utilized for the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • Targets somatostatin receptor type 2 (SST2)
  • Octreotide derivative
  • Used for synthesis/research of radionuclide-drug conjugates (RDCs)
Catalog No. 50-004-24987 Supplier Medchemexpress LLC Supplier No. HYP107585MG
Alkyne-βAG-TOCA is an octreotide derivative that targets somatostatin receptor type 2 (SST2). This compound is utilized for the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • Targets somatostatin receptor type 2 (SST2)
  • Octreotide derivative
  • Used for synthesis/research of radionuclide-drug conjugates (RDCs)